Learn More
Abnova™ Chloride Assay Kit
To quantify chloride, functional study
Manufacturer:  Abnova™ KA0823
Catalog No. 89-028-076
Add to cart
Includes:
  • Chloride reagent, 15mL
  • Chloride standard (10µmol), lyophilized, 1 vial
Specifications
Specifications
20 to 120nmol
Functional Study
1 Kit
Store the kit at room temperature.
Chloride Assay Kit
Chloride Assay Kit is used for quantifying chloride.
20mL Chloride reagent, 1 vial Chloride standard (10μM)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric