Learn More
Abnova™ Cholesterol/Cholesteryl Ester Quantitation Kit
Used for detecting total cholesterol (cholesterol and cholesteryl esters) in the presence of cholesterol esterase, or free cholesterol in the absence of cholesterol esterase in the reaction
Manufacturer:  Abnova™ KA0829
Catalog No. 89-028-071
Add to cart
Description
Description
  • Sample Type: Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine
Functional Study
Specifications
Specifications
Cholesterol/Cholesteryl Ester Quantitation Kit is used for detecting total cholesterol (cholesterol and cholesteryl esters) in the presence of cholesterol esterase, or free cholesterol in the absence of cholesterol esterase in the reaction.
25mL Cholesterol assay buffer, 200μL Cholesterol probe (in DMSO, anhydrous), 1 vial enzyme mix (lyophilized), 1 vial Cholesterol Esterase (lyophilized), 100μL Cholesterol standard (2μg/μ
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric, Fluorometric
Functional Study
1 Kit
Store the kit at -20°C.
Cholesterol/Cholesteryl Ester Quantitation Kit