Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Choline/Acetylcholine Quantification Kit

To quantifying choline and acetylcholine, functional study

Manufacturer:  Abnova™ KA0840

Catalog No. 89-028-064


Add to cart

Includes:
  • Choline assay buffer, 25mL
  • Choline probe, 200µL
  • Choline enzyme mix, 1 vial
  • Acetylcholinesterase, 1 vial
  • Choline standard (5µmol), 1 vial

Specifications

Specifications

10pM to 5nM
Functional Study
1 Kit
Store the kit at -20°C.
Choline/Acetylcholine Quantification Kit
Choline/Acetylcholine Quantification Kit is used for quantifying choline and acetylcholine by either a colorimetric or fluorometric method.
25mL Choline assay buffer, 200μL Choline probe, 1 vial Choline enzyme mix, 1 vial Acetylcholinesterase, 1 vial Choline standard (5μM)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric, Fluorometric
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit