Learn More
Abnova™ Citrate Assay Kit
Used for quantifying citrate and functional study
Manufacturer:  Abnova™ KA0864
Catalog No. 89-024-419
Add to cart
Specifications
Specifications
0.1 to 10nm
Functional Study
1 Kit
Store the kit at -20°C.
Citrate Assay Kit
Citrate Assay Kit is used for quantifying citrate.
25mL Citrate assay buffer, 0.2mL Citrate probe, 1 vial Citrate enzyme mix (lyophilized), 1 vial Citrate developer (lyophilized), 1 vial Citrate standard (10μM, lyophilized)
Biological fluid, cell culture supernatant, tissue culture supernatant, urine
Colorimetric, Fluorometric