Learn More
Abnova™ Coenzyme A Assay Kit
To measure the CoA level, functional study
Manufacturer:  Abnova™ KA0809
Catalog No. 89-028-089
Add to cart
Specifications
Specifications
2.5 to 250μM
Functional Study
1 Kit
Store the kit at -20°C.
CoA Assay Kit
Coenzyme A Assay Kit is used for measuring the CoA level.
25mL CoA assay buffer, 200μL OxiRed™ probe, 1 vial conversion enzyme mix, 1mL CoA substrate, 1 vial Acyl CoA developer, 1 vial CoA standard
Biological fluid, cell lysate, culture medium, plasma, serum, tissue lysate, urine
Colorimetric, Fluorometric