Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cortisol (Human) ELISA Kit

Used for the quantitative measurement of Cortisol in human serum or plasma.

Manufacturer:  Abnova™ KA0918

Catalog No. 89-024-459

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Includes:
  • Microwells coated with Cortisol MAb 12x8x1
  • Cortisol Standard: 7 vials (Ready to use) 0.7mL
  • Enzyme Conjugate (20X) 1.2mL
  • TMB Substrate: 1 bottle (Ready to use) 12mL
  • Stop Solution: 1 bottle (Ready to use) 12mL
  • 20X Wash concentrate: 1 bottle 25mL
  • Assay Diluent 24mL

Description

Description

Solid-phase competitive ELISA
Specifications

Specifications

20 to 800ng/mL
Human
Colorimetric
12 x 8 x 1 Microwell coated with Cortisol MAb, 0.7mL Cortisol standard, 1.2mL (20X) Enzyme Conjugate, 12mL TMB Substrate, 12mL stop solution, 25mL (20X) Wash concentrate, 24mL Assay diluent
96 Assays
4°C
HRP
1.5ng/mL
For the quantitative measurement of Cortisol in Human Serum or Plasma
Cortisol (Human) ELISA kit
Plasma, Serum
25μL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit