Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cortisol (Sheep) ELISA Kit

Immunoassay for the quantitative determination of cortisol in sheep serum, plasma, urine

Manufacturer:  Abnova™ KA0919

 View more versions of this product

Catalog No. 89-024-460


Due to product restrictions, please to purchase or view availability for this product.


Specifications

Specifications

1 to 80ng/mL
Sheep
0.1ng/mL
Quantification
Cortisol (Sheep) ELISA Kit
RUO
Store the kit at 4°C.
HRP
Cortisol (Sheep) ELISA Kit is used for the quantitative measurement of Cortisol in sheep serum or plasma.
Colorimetric
12 x 8 x 1 Microwell coated with Cortisol MAb, 0.7mL Cortisol standard, 1.2mL (20X) Enzyme Conjugate, 12mL TMB Substrate, 12mL stop solution, 25mL (20X) Wash concentrate, 24mL Assay diluent
96 Assays
Plasma, Serum
40μL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit