Learn More
Abnova™ Creatine Assay Kit
Used for measuring creatine and functional study
Manufacturer:  Abnova™ KA0856
Catalog No. 89-024-411
Add to cart
Specifications
Specifications
0.001 to 10mm
Functional Study
1 Kit
Store the kit at -20°C.
Creatine Assay Kit
Creatine Assay Kit is used for measuring creatine.
25mL Creatine assay buffer, 0.2mL Creatine probe (in DMSO), 1 vial Creatinase (lyophilized), 1 vial Creatine enzyme mix (lyophilized), 1 vial Creatine standard (10μM, lyophilized)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric, Fluorometric