Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Creatinine Assay Kit

Used for measuring creatine concentration and functional study

Manufacturer:  Abnova™ KA0849

Catalog No. 89-024-404


Add to cart

Includes:
  • Creatinine assay buffer, 25mL
  • Creatinine probe, 0.2mL
  • Creatinase, 1 vial
  • Creatininase (lyophilized), 1 vial
  • Creatinine enzyme mix (lyophilized), 1 vial
  • Creatinine (10µmol), lyophilized, 1 vial

Specifications

Specifications

Creatinine Assay Kit is used for measuring creatine concentration.
25mL Creatinine assay buffer, 0.2mL Creatinine probe, 1 vial Creatinase, 1 vial Creatininase (lyophilized), 1 vial Creatinine enzyme mix (lyophilized), 1 vial Creatinine (10μM) lyophilized
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric, Fluorometric
Functional Study
1 Kit
Store the kit at -20°C.
Creatinine Assay Kit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit