Learn More
Abnova™ CXCL1 (Human) ELISA Kit
Manufacturer:  Abnova Corporation KA1722
 View more versions of this product
Catalog No. 89-028-458
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Biotin
50pg/mL
For the quantitative measurement of Human CXCL1
CXCL1 (Human) ELISA kit
Cell Culture Supernatant, Plasma, Serum, Urine
4°C
Human
Colorimetric
96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP Streptavidian concentrate, 12mL TMB One-Step Substrate reagent, 8mL stop solut
96 Assays
100μL
100μL