Learn More
Abnova™ Cytochrome p450 Gene Knockdown Quantification Kit
Cytochrome p450 Gene Knockdown Quantification Kit is used for measuring cytochrome p450 knockdown induced by siRNA or antisense oligonucleotide at the protein level.
Manufacturer:  Abnova™ KA1546
Catalog No. 89-028-240
Add to cart
Description
Description
  • Quick and efficient procedure; completion of entire assay needs only 4 hours
  • Innovative colorimetric assay without the need for radioactivity, electrophoresis, or chromatography
  • Internal control is conveniently included to correct for the variations for the cell number or protein concentrations
  • Strip microplate format makes the assay flexible, manual or high throughput
  • Simple, reliable, and consistent assay conditions
  • Kit is shipped in two parts, one at ambient room temperature and the second frozen on ice packs at 4°C
Specifications
Specifications
Cytochrome p450 Gene Knockdown Quantification Kit is used for measuring cytochrome p450 knockdown induced by siRNA or antisense oligonucleotide at the protein level.
Each
10μL
Cyto chrome p450 gene knockdown quantification kit
Quantification
Store the detection antibody at -20°C. Store Q2, Q4, Q6, GAPDH control antibody and 8-well assay strips at 4°C away from light. Store all other components at room temperature.