Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ D-Lactate Assay Kit (Colorimetric)
Used for measuring D-lactate and functional study
Supplier: Abnova™ KA0869
Specifications
0.01 to 10mm | |
D-Lactate Assay Kit (Colorimetric) is used for measuring D-lactate. | |
Functional Study | |
1 Kit | |
Cellular Assay Kit |
Store the kit at -20°C. | |
Colorimetric | |
25mL D-Lactate assay buffer, 1 vial D-Lactate enzyme mix (lyophilized), 1 vial D-Lactate substrate mix (lyophilized), 100μL D-Lactate standard (100mM) | |
Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine, Fermentation Medium |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction