Learn More
Abnova™ Dengue virus IgM ELISA Kit
For detection of IgM antibody to Dengue Virus in Human Serum or Plasma
Manufacturer:  Abnova™ KA0962
Catalog No. 89-028-057
Add to cart
Specifications
Specifications
HRP
Colorimetric
12 x 8 x 1 Microwell, 22mL sample diluent, 1.5mL Calibrator, 1.5mL positive control, 1.5mL negative control, 12mL Enzyme Conjugate, 12mL TMB Chromogenic Substrate, 12mL stop solution, 20 x 20mL washing concentrate
96 Assays
Store the kit at 4°C.
Dengue virus IgM ELISA Kit is used for the detection of IgM antibody to Dengue virus in human serum or plasma.
Qualitative
Dengue virus IgM ELISA Kit
Plasma, Serum
10μL