Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Dihydrotestosterone ELISA Kit

Enzyme immunoassay for the quantitative determination of Dihydrotestosterone in human serum

Manufacturer:  Abnova™ KA1886

Catalog No. 89-028-619


Add to cart

Description

Description

  • Sample Type: Serum
  • Sample Volume: 50µL

Quantification

Specifications

Specifications

Dihydrotestosterone ELISA Kit is an enzyme immunoassay for the quantitative determination of Dihydrotestosterone in human serum.
Quantification
Dihydrotestosterone ELISA Kit
1 Kit
Store the kit at 4°C.
Colorimetric
96-microwell Rabbit Anti-DHT Antibody coated, 0.2mL Dihydrotestosterone-HRP (HRP) Conjugate (100X), 2mL Dihydrotestosterone standards A, 0.6mL Dihydrotestosterone standards B,, 0.6mL Dihydrotestosterone standards C, 0.6mL Dihydrotestosterone standards D,
Dihydrotestosterone ELISA kit
Serum
50μL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit