Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ DNA Damage Quantification Kit
To determine the DNA lesion and repair, functional study
Supplier: Abnova™ KA0784
Specifications
Store the kit at 4°C. Do not freeze. | |
Colorimetric | |
0.125mL ARP solution (10mM, in DMSO), 30mL TE buffer, 0.1mL glycogen solution (10μg/μL), 0.6mL 0 ARP-DNA standard (0.5μg/mL), 0.6mL 40 ARP-DNA standard (0.5μg/mL) | |
Quantification Kit |
DNA Damage Quantification Kit is used for determining the DNA lesion and repair. | |
Functional Study | |
1 Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction