Learn More
Abnova™ DNA Methyltransferase Activity/Inhibition Assay Kit (Colorimetric)
For measuring Dnmt activity/inhibition
Manufacturer:  Abnova™ KA0680
Catalog No. 89-027-037
Add to cart
Specifications
Specifications
DNA Methyltransferase Activity/Inhibition Assay Kit (Colorimetric) is use for measuring Dnmt activity/inhibition.
Functional Study
DNA Methyltransferase Activity/Inhibition Assay Kit (Colorimetric)
1 Kit
Colorimetric
22mL Wash Buffer (10X) (M1), 3mL DNMT Assay Buffer (M2), 70μL adomet (M3), 10μL DNMT positive control (M4), 8μL capture Antibody (M5), 20μL detection Antibody (M6), 12mL developing so
DNA Methyltransferase activity/inhibition assay kit
Store M3, M4, and M6, at -20°C away from light. Store all other components at 4°C away from light.