Learn More
Abnova™ DNMT Activity/Inhibition Assay Ultra Kit (Colorimetric)
To screen DNMT inhibitors, functional study
Manufacturer:  Abnova™ KA1515
Catalog No. 89-028-210
Add to cart
Includes:
  • MU1 (10X wash buffer), 14mL
  • MU2 (DNMT assay buffer), 4mL
  • MU3 (Adomet, 50x), 60µL
  • MU4 (DNMT enzyme control, 50µg/mL), 6µL
  • MU5 (capture antibody, 1000µg/mL), 5µL
  • MU6 (detection antibody, 400µg/mL), 6µL
  • MU7 (enhancer solution), 6µL
  • MU8 (developer solution), 5mL
  • MU9 (stop solution), 5mL
  • Six 8-Well assay strips (with frame)
  • Adhesive covering film
Specifications
Specifications
0.5 to 20μg nuclear extract, 0.5 to 200ng purified Enzyme
Functional Study
1 Kit
1 to 5μL
Colorimetric
DNMT Activity/Inhibition Assay Ultra Kit (Colorimetric) is use for screening DNMT inhibitors.
14mL MU1 (wash buffer 10X), 4mL MU2 (DNMT assay buffer), 60μL MU3 (Adomet, 50x), 6μL MU4 (DNMT enzyme control, 50μg/mL), 5μL MU5 (Capture antibody, 1000μg/mL), 6
Nuclear extract, purified enzyme
Store MU3, MU4, MU6 and MU7 at -20°C away from light. Store MU1, MU5, MU8 and 8-Well Assay Strips at 4°C away from light. Store all other components at room temperature away from light. Aliquot to avoid repeated freezing and thawing.
DNMT Activity/Inhibition Assay Ultra Kit