Learn More
Abnova™ Elastase Ab ELISA Kit
Indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against Elastase in Human Serum or Plasma
Manufacturer:  Abnova™ KA1102
Catalog No. 89-027-240
Add to cart
Specifications
Specifications
6.3 to 100U/mL
Elastase Ab ELISA Kit is an indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against Elastase in human serum or plasma.
Quantification
Elastase Ab ELISA Kit
Plasma, Serum
10μL
HRP
Colorimetric
6 1.5mL combined calibrator with IgG class Anti-elastase Antibody vials, 2 1.5mL Anti-elastase control vials, one 20mL Sample Buffer vial, one 15mL Enzyme Conjugate solution vial, one 15mL TMB Substrate solution vial, one 15mL stop solution vial, one 96-well plate, one 20mL wash solution vial
96 Tests
Store the kit at 4°C.