Learn More
Abnova™ ENA-4 Profile ELISA Kit
Immunoassay, ELISA, ENA-4 profile ELISA kit, HRP, Indirect solid phase ELISA for semi-quantitative determination of IgG autoabs against extractable nuclear antigens SS-A (Ro), SS-B (La), Sm and RNP/Sm in Human Serum, Plasma
Manufacturer:  Abnova™ KA1104
Catalog No. 89-027-242
Add to cart
Specifications
Specifications
HRP
Colorimetric
1 x 2mL Positive control vial, 1x 2mL negative control vial, 4 x 1mL Cut-off calibrator vial, one 20mL Sample Buffer, one 15mL Enzyme Conjugate solution
96 Tests
Store the kit at 4°C.
ENA-4-Profile ELISA Kit is an indirect solid phase ELISA for semi-quantitative determination of IgG class autoantibodies directed against the extractable nuclear antigens SS-A (Ro), SS-B (La), Sm and RNP/Sm in human serum or plasma.
Quantification
ENA-4 Profile ELISA Kit
Plasma, Serum
10μL