Learn More
Abnova™ Europium Calibration Kit
Used for optimizing and calibrating the europium signal output in all time-resolved fluorescence readers
Manufacturer:  Abnova™ KA2097
Catalog No. 89-028-729
Add to cart
Specifications
Specifications
Europium Calibration Kit is used for optimizing and calibrating the europium signal output in all time-resolved fluorescence readers.
50mL Enhancement solution, Europium Chloride 100nM
Store at 4°C.
Control
1 Kit
Europium Calibration Kit