Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Europium Labeling Kit
Manufacturer: Abnova™ KA2096
Includes:
- Lyophilized W1024 chelate(140nmol), 100μg
- Europium standard, 0.5mL
- Enhancement solution, 50mL
- 96-Well plate, 1 plate
Description
Europium Labeling Kit is used for simple and rapid preparation of Europium-labeled antibodies and proteins
Conjugation
Specifications
Europium Labeling Kit | |
Conjugation | |
140nmol 100μg lyophilized W1024 chelate, 0.5mL europium standard, 50mL enhancement solution, one 96 well plate | |
1 Kit |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title