Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ F7 (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human F7
$1192.00
Specifications
Conjugate | Biotin |
---|---|
Cross Reactivity | Human |
Description | F7 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human F7. |
Detection Method | Colorimetric |
Includes | 96 Antibody coated microwell, 270ng Human FVII standard, Biotinylated FVII Antibody(100X) vial, 30mL MIX diluent concentrate (10X), 2 30mL Wash Buffer 20X, 80 Streptavidin-Peroxidase Conjugate, 8mL Chromogen Substrate, 12mL stop solution |
Catalog Number | Mfr. No. | Calibration Range | Detection Limit | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Calibration Range | Detection Limit | Content And Storage | Price | Quantity & Availability | |||
89-028-590
|
Abnova™
KA1837 |
0.352 to 90ng/mL | 0.3ng/mL | Store components of the kit at 4°C or -20°C as described in the protocol. |
Each for $1,192.00
|
|
|||
89-100-316
|
Abnova™
KA0471 |
6.25 to 200ng/mL | 6ng/mL | Store components of the kit at 4°C or -20°C as described in the protocol. |
Each for $1,192.00
|
|
|||
Specifications
Biotin | |
F7 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human F7. | |
96 Antibody coated microwell, 270ng Human FVII standard, Biotinylated FVII Antibody(100X) vial, 30mL MIX diluent concentrate (10X), 2 30mL Wash Buffer 20X, 80 Streptavidin-Peroxidase Conjugate, 8mL Chromogen Substrate, 12mL stop solution | |
For research use only (RUO) | |
50 μL | |
Quantification | |
96 |
Human | |
Colorimetric | |
1 Kit | |
Cell Culture Supernatant, Plasma, Serum | |
Store components of the kit at 4°C or -20°C as described in the protocol. | |
F7 (Human) ELISA kit |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title