Learn More
Abnova™ FAD Assay Kit
To provide a rapid, simple, ultra-sensitive and reliable test suitable for high throughput assay of FAD, functional study
Manufacturer:  Abnova™ KA0808
Catalog No. 89-028-090
Add to cart
Specifications
Specifications
FAD Assay Kit is used for providing a rapid, simple, ultra-sensitive, and reliable test suitable for high throughput assay of FAD.
25mL FAD assay buffer, 0.2mL OxiRed™ probe, 1 vial FAD enzyme mix, 1 vial FAD standard, 1 vial stop solution
Biological fluid, cell lysate, culture medium, plasma, serum, tissue lysate, urine
Colorimetric, Fluorometric
Functional Study
1 Kit
Store the kit at -20°C.
FAD Assay Kit