Learn More
Abnova™ FastLink Alkaline Phosphatase Labeling Kit
Manufacturer:  Abnova Corporation KA1549
Catalog No. 89-028-243
Add to cart
Description
Description
FastLink Alkaline Phosphatase Labeling Kit is used for simple and rapid preparation of alkaline phosphatase-labeled antibodies and proteins
Conjugation
Specifications
Specifications
Conjugation, used for simple and rapid preparation of alkaline phosphatase labeled antibodies and proteins
FastLink Alkaline Phosphatase Labeling Kit
-20°C
Lyophilized
96 well FastLink mix, vial modifier reagent, vial quencher reagent
3 x 10, 3 x 100μg/KT
Brown
12 Months