Learn More
Abnova™ FastLink Biotin Labeling Kit
Manufacturer:  Abnova Corporation KA1576
Catalog No. 89-028-268
Add to cart
Description
Description
FastLink Biotin Labeling Kit is used for simple and rapid preparation of biotin-labeled antibodies and proteins This kit optimizes for assays in which the biotinylated protein is captured by streptavidin immobilized on a surface (ie plates, nitrocellulose, magnetic beads etc)
Conjugation
Specifications
Specifications
Conjugation, used for simple and rapid preparation of biotin-labeled antibodies and proteins
FastLink Biotin Labeling Kit
-20°C
Lyophilized
96 well FastLink mix, vial modifier reagent, vial quencher reagent
3 x 20, 3 x 200μg/KT
Brown
12 Months