Learn More
Abnova™ FastLink Biotin Labeling Kit
Manufacturer:  Abnova Corporation KA1575
Catalog No. 89-028-267
Add to cart
Description
Description
FastLink Biotin Labeling Kit is used for simple and rapid preparation of biotin-labeled antibodies and proteins This kit optimizes for assays in which a streptavidin-labeled reagent will be used
Conjugation
Specifications
Specifications
Conjugation, used for simple and rapid preparation of biotin-labeled antibodies and proteins
FastLink Biotin Labeling Kit
-20°C
Lyophilized
96 well FastLink mix, vial modifier reagent, vial quencher reagent
3 x 20, 3 x 200μg/KT
Brown
12 Months