Learn More
Abnova™ FastLink Fluorescein Labeling Kit
Manufacturer:  Abnova Corporation KA1571
Catalog No. 89-028-263
Add to cart
Description
Description
FastLink Fluorescein Labeling Kit is used for simple and rapid preparation of Fluorescein labeled antibodies and proteins
Conjugation
Specifications
Specifications
Conjugation, used for simple and rapid preparation of fluorescein labeled antibodies and proteins
FastLink Fluorescein Labeling Kit
-20°C
Lyophilized
96 well FastLink mix, vial modifier reagent, vial quencher reagent
3 x 20, 3 x 200μg/KT
Yellow
12 Months