Learn More
Abnova™ FastLink Glucose Oxidase Labeling Kit
Manufacturer:  Abnova Corporation KA1550
Catalog No. 89-028-244
Add to cart
Description
Description
FastLink Glucose Oxidase Labeling Kit is used for simple and rapid preparation of glucose oxidase-labeled antibodies and proteins
Conjugation
Specifications
Specifications
Conjugation, used for simple and rapid preparation of glucose oxidase labeled antibodies and proteins
FastLink Glucose Oxidase Labeling Kit
-20°C
Lyophilized
96 well FastLink mix, vial modifier reagent, vial quencher reagent
3 x 10, 3 x 100μg/KT
Brown
12 Months