Learn More
Abnova™ FastLink PE-Texas Red Tandem Labeling Kit
Manufacturer:  Abnova Corporation KA1582
Catalog No. 89-028-270
Add to cart
Description
Description
FastLink PE-Texas Red Tandem Labeling Kit is used for simple and rapid preparation of PE-Texas Red Tandem-labeled antibodies and proteins
Conjugation
Specifications
Specifications
Conjugation, used for simple and rapid preparation of PE-Texas red tandem-labeled antibodies and proteins
FastLink PE-Texas Red Tandem Labeling Kit
-20°C
Lyophilized
96 well FastLink mix, vial modifier reagent, vial quencher reagent
3 x 10, 3 x 100μg/KT
Brown
12 Months