Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FastLink PerCP Labeling Kit

For simple and rapid preparation of PerCP labeled antibodies and proteins

Manufacturer:  Abnova Corporation KA1552

Catalog No. 89-139-156

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Three glass vial(s) of FastLink mix (depending on pack size), one vial of Modifier reagent, one vial of Quencher reagent

Description

Description

  • Fluorophore: Peridinin Chlorophyll Protein (PerCP)
  • Reactive Group: Amine group
  • Excitation: 482nm
  • Emission: 677nm
  • Application: Conjugation

  • Storage: Store at -20°C.

  • Specifications

    Specifications

    Conjugation, used for simple and rapid preparation of PerCP labeled antibodies and proteins
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit

    For research use only (RUO).