Learn More
Abnova™ FastLink R-PE Labeling Kit
Manufacturer:  Abnova Corporation KA1553
Catalog No. 89-028-247
Add to cart
Includes: 3 x 10µg/Kit
  • FastLink mix, Glass vial
  • Modifier reagent, 1 vial
  • Quencher reagent, 1 vial
3 x 100µg/Kit
  • FastLink mix, 3 Glass vial
  • Modifier reagent, 1 vial
  • Quencher reagent, 1vial
Description
Description
FastLink R-PE Labeling Kit is used for simple and rapid preparation of R-PE labeled antibodies and proteins
Conjugation
Specifications
Specifications
Conjugation, used for simple and rapid preparation of R-PE labeled antibodies and proteins
FastLink R-PE Labeling Kit
-20°C
Lyophilized
96 well FastLink mix, vial modifier reagent, vial quencher reagent
3 x 10, 3 x 100μg/KT
Brown
12 Months