Learn More
Abnova™ FastLink Streptavidin Labeling Kit
Manufacturer:  Abnova Corporation KA1556
Catalog No. 89-028-250
Add to cart
Description
Description
FastLink Streptavidin Labeling Kit is used for simple and rapid preparation of streptavidin labeled antibodies and proteins
Conjugation
Specifications
Specifications
Conjugation, used for simple and rapid preparation of streptavidin labeled antibodies and proteins
FastLink Streptavidin Labeling Kit
-20°C
Lyophilized
96 well FastLink mix, vial modifier reagent, vial quencher reagent
3 x 10, 3 x 100μg/KT
Brown
12 Months