Learn More
Abnova™ Fentanyl (Human) ELISA Kit
Sensitive in-vitro test to detect the presence of Fentanyl in samples such as whole Blood, Serum, Plasma and Urine
Manufacturer:  Abnova™ KA0931
Catalog No. 89-024-472
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
0.5 to 5ng/mL
Human
Colorimetric
96-microwell strip, 12mL Enzyme Conjugate, 2mL positive reference standard, 1mL negative control, 12mL TMB chromogenic Substrate, 11mL stop solution
96 Assays
Store the kit at 4°C.
HRP
Fentanyl (Human) ELISA Kit is a sensitive in-vitro test to detect the presence of Fentanyl in samples such as whole blood, serum, plasma and urine.
Qualitative
Fentanyl (Human) ELISA Kit
Plasma, Serum, Urine, Whole Blood
20μL