Learn More
Abnova™ Ferric Reducing Ascorbate Assay Kit
Used for detecting ascorbic acid and functional study
Manufacturer:  Abnova™ KA0870
Catalog No. 89-024-425
Add to cart
Specifications
Specifications
0.2 to 20nm
Functional Study
1 Kit
Store the kit at -20°C.
Ferric reducing Ascorbate Assay Kit
Ferric Reducing Ascorbate Assay Kit is used for detecting ascorbic acid.
25mL FRASC buffer, 1mL Ascorbic Acid probe, 1mL FeCl3 solution, 1 vial Ascorbate Oxidase (lyophilized), 1 vial Ascorbic Acid standard (20μM)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine, food sample, growth medium
Colorimetric