Learn More
Abnova™ Fibrinogen (Dog) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Dog Fibrinogen in Plasma
Manufacturer:  Abnova™ KA1402
Catalog No. 89-024-522
Add to cart
Specifications
Specifications
1 to 10μg/mL
Dog
Colorimetric
96 Antibody coated microwell, 20μg Dog FBG standard, Biotinylated FBG, 30mL EIA diluent (10X), 30mL Wash Buffer (20X), 80μL Streptavidin Peroxidase Conjugate, 8mL Chromogen Substrate, 12mL stop solution
Fibrinogen (Dog) ELISA kit
Plasma
25μL
Biotin
Fibrinogen (Dog) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of dog fibrinogen in plasma.
Quantification
Fibrinogen (Dog) ELISA Kit
1 Kit
Store the kit at 4°C or -20°C.