Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 630-N NHS Ester

Water insoluble fluorescent dye derived from dipyrrometheneboron difluoride

Manufacturer:  Abnova™ U0148

Catalog No. 89-139-434

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Chemical Formula: C44H47BF2N6O6
  • Mol. Weight: 804.69g/mol
  • Purity: ≥98%
  • Molar Extinction Co-efficient: 1,13,000 M-1cm-1 (CH3COH)
  • Excitation (Max): 629nm (CH3OH)
  • Emission (Max): 646nm (CH3OH)
  • Recommended Excitation: 632nm red HeNe and 635nm red diode
  • Solubility: Methanol, DMSO, DMF, acetonitrile, dichloromethane

Application: Conjugation

Recommended Storage: Store in the dark at -20°C.

Specifications

Specifications

Fluorescent dye derived from dipyrrometheneboron difluoride. Water insoluble and forms stable amide bond with primary amines at pH 7-9.
Conjugation
7 to 9
Store in the dark at -20°C.
804.69g/mol
646nm (CH3OH)
≥98%
Derived from dipyrrometheneboron difluoride
Fluorescent Dye 630-N NHS Ester
Fluorescent Dye 630-N NHS Ester
5 mg
629nm (CH3OH)
632nm red HeNe and 635nm red diode
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only (RUO).