Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 678-Z Acid

Water insoluble fluorescent dye derived from Cyanine

Manufacturer:  Abnova™ U0169

Catalog No. 89-139-455

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Chemical Name: 2-((1E,3E,5E)-5-(3-(5-carboxypentyl)-1,1-dimethyl-1H-benzo[e]indol-2(3H)-ylidene)penta-1,3-dienyl)-3-ethyl-1,1-dimethyl-1H-benzo[e]indolium-6-sulfonate
  • Chemical Formula: C41H44N2O5S
  • Mol. Weight: 676.86g/mol
  • Purity: ≥95%
  • Molar Extinction Co-efficient: 1,95,000 M-1cm-1 (CH3COH)
  • Excitation (Max): 678nm (CH3OH)
  • Emission (Max): 705nm (CH3OH)
  • Recommended Excitation: Argon-Krypton ion laser 647nm, Red diode laser 650nm, 670nm
  • Recommended Filter Sets: CyR5.5, LI-COR 700 channel
  • Solubility: Acetonitrile, Dichloromethane, DMF, DMSO, Methanol

Application: Conjugation

Recommended Storage: Store in the dark at 4°C and keep under argon.

Specifications

Specifications

Fluorescent dye derived from Cyanine. Water insoluble and forms stable amide bond with primary amines via carbodiimide activation.
678 (CH3OH)
Derived from Cyanine
5 mg
676.86g/mol
CyR5.5, LI-COR 700 channel
705 (CH3OH)
Fluorescent Dye 678-Z Acid
Conjugation
Fluorescent Dye 678-Z Acid
Argon-Krypton ion laser 647nm, Red diode laser 650nm, 670nm
≥95%
Safety and Handling

Safety and Handling

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only (RUO).