Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 830-C NHS Ester

Water insoluble fluorescent dye derived from Cyanine

Manufacturer:  Abnova™ U0189

Catalog No. 89-139-475

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Chemical Name: 2-((E)-2-((E)-2-(4-(5-(2,5-dioxopyrrolidin-1-yloxy)-5-oxopentanamido)phenylthio)-3-((Z)-2-(3-ethyl-1,1-dimethyl-1H-benzo[e]indol-2(3H)-ylidene)ethylidene) cyclohex-1-enyl)vinyl)-3-ethyl-1,1-dimethyl-1H-benzo[e]indolium iodide
  • Chemical Formula: C57H59IN4O5S
  • Mol. Weight: 1039.07g/mol
  • Purity: ≥95%
  • Molar Extinction Co-efficient: 2,50,000 M-1cm-1 (CH3COH)
  • Excitation (Max): 827nm (CH3OH)
  • Emission (Max): 867nm (CH3OH)
  • Recommended Excitation: Dye-pumped laser, Xenon-Arc lamp
  • Solubility: DMF, DMSO, Methanol, Acetonitrile, Dichloromethane

Application: Conjugation

Recommended Storage: Store in the dark at -20°C and keep under argon.

Specifications

Specifications

Fluorescent dye derived from Cyanine. Water insoluble and forms stable amide bond with primary amines at pH 7-9.
Conjugation
7 to 9
Store in the dark at -20°C and keep under argon.
1039.07g/mol
867nm (CH3OH)
≥95%
Derived from Cyanine
Fluorescent Dye 830-C NHS Ester
Fluorescent Dye 830-C NHS Ester
5 mg
827nm (CH3OH)
Dye-pumped laser, Xenon-Arc lamp
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only (RUO).