Learn More
Abnova™ Fluorescent In Situ Hybridization (FISH) Buffer
Optimized for performance with FISH probes
Manufacturer:  Abnova™ U0029
 View more versions of this product
Catalog No. 89-028-828
Add to cart
Description
Description
Fluorescent In Situ Hybridization (Cell)
Specifications
Specifications
FISH Hybridization Buffer is optimized for performance with FISH probes and contains dextran sulfate, formamide, and SSC (pH 7.0).
FISH Hybridization Buffer(5ml)
Store at 4°C
Fluorescent In Situ Hybridization (Cell)
5 x 1 mL