Learn More
Abnova™ Fluorescent Phosphate Assay Kit
To assess phosphate over a wide range of concentrations, functional study
Manufacturer:  Abnova™ KA0818
Catalog No. 89-028-081
Add to cart
Specifications
Specifications
0.4 to 10μM
Functional Study
1 Kit
Store the kit at 4°C.
Fluorescent Phosphate Assay Kit
Fluorescent Phosphate Assay Kit is used for assessing phosphate over a wide range of concentrations.
25mL Phosphate assay buffer, 1 vial OxiRed™ probe, 0.4mL Dimethylsulfoxide (DMSO dried), 1 vial Maltose converter, 1 vial Glucose developer, 0.2mL substrate, 50μL Phosphate standard (100mM)
Biological fluid, cell lysate, culture medium, plasma, serum, tissue lysate, urine
Fluorometric