Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Free Glycerol Assay Kit
Used for measuring free glycerol concentration and functional study
Supplier: Abnova™ KA0853
Specifications
1 to 10000μM | |
Free Glycerol Assay Kit is used for measuring free glycerol concentration. | |
Functional Study | |
1 Kit | |
Detection Assay Kit |
Store the kit at -20°C. | |
Colorimetric, Fluorometric | |
25mL Glycerol assay buffer, 0.2mL Glycerol probe (in DMSO, anhydrous), 1 vial Glycerol enzyme mix (lyophilized), 0.2mL Glycerol standard (100mM) | |
Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction