Learn More
Abnova™ Fumarate Assay Kit
Used for detecting fumarate and functional study
Manufacturer:  Abnova™ KA0855
Catalog No. 89-024-410
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
Fumarate Assay Kit is used for detecting fumarate.
25mL Fumarate assay buffer, 1 vial Fumarate enzyme mix, 1 vial Fumarate developer, 0.2mL Fumarate standard (0.1M)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric
Functional Study
1 Kit
Store the kit at -20°C.
Fumarate Assay Kit