Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Galactose and Lactose Assay Kit
To measure galactose and lactose levels, functional study
Supplier: Abnova™ KA0842
Includes:
- Galactose assay buffer, 25mL
- Galactose probe (DMSO solution), 0.2mL
- Lactase (lyophilized), 1 vial
- Galactose enzyme mix (lyophilized), 1 vial
- HRP (lyophilized), 1 vial
- Galactose standard (100nmol/μL) 10μL
Specifications
Store the kit at -20°C. | |
Colorimetric, Fluorometric | |
25mL Galactose assay buffer, 0.2mL Galactose probe (DMSO solution), 1 vial Lactase (lyophilized), 1 vial Galactose enzyme mix (lyophilized), 1 vial HRP (lyophilized), 100μL Galactose standard (100nM/μL) | |
Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine |
Galactose and Lactose Assay Kit is used for measuring of galactose and lactose levels. | |
Functional Study | |
100 Assays | |
Activity Detection Assay Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction