Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ GFP Quantitation Kit

GFP Quantitation Kit is used for quantifying GFP.

Manufacturer:  Abnova™ KA0911

Catalog No. 89-024-456


Due to product restrictions, please to purchase or view availability for this product.


Includes:
  • GFP Assay Buffer: 25mL
  • GFP Standard (1µg/µL): 100µL
  • GFP Quench Solution: 1mL

Description

Description

  • GFP and EGFP have been widely used as a valuable reporter molecule for in vivo visualization of gene expression events in a variety of cell types and organisms
  • 96 microplate format
  • Cells or tissues can be homogenized directly in the GFP Assay Buffer
  • Kit can detect a wide range of GFP concentration (o.01 to 10 µg/mL)
  • Store kit at -20°C

Specifications

Specifications

GFP Quantitation Kit is used for quantifying GFP.
GFP assay buffer 25 mL
Store the kit at -20°C
100 Assays
Functional Study
GFP Quantitation Kit
100 Assays
Cell, Tissue
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit