Learn More
Abnova™ Gliadin IgA ELISA Kit
Indirect solid phase Enzyme Immunoassay for the quantitative measurement of specific IgA antibodies against Gliadin in Serum and Plasma
Manufacturer:  Abnova™ KA1286
Catalog No. 89-024-578
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
HRP
Gliadin IgA Ab ELISA Kit is an indirect solid phase enzyme immunoassay for the quantitative measurement of specific IgA antibodies against Gliadin in serum and plasma.
Quantification
Gliadin IgA ELISA Kit
1 Kit
Store the kit at 4°C.
Human
Colorimetric
12 Gliadin Antigen coated microtiter strip, 2mL calibrator A (negative control), 2mL calibrator B (cut-off standard), 2mL calibrator C (weak positive control), 2mL calibrator D (positive control), 15mL Enzyme Conjugate, 15mL Substrate, 15mL stop solution, 60mL sample diluent, 60mL Wash Buffer (10X)
Gliadin IgA ELISA kit
Plasma, Serum
100μL