Learn More
Abnova™ Gliadin IgM ELISA Kit
Indirect solid phase Enzyme Immunoassay for the quantitative measurement of specific IgM antibodies against Gliadin in Serum and Plasma
Manufacturer:  Abnova™ KA1287
Catalog No. 89-024-579
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
HRP
Gliadin IgM Ab ELISA Kit is an indirect solid phase enzyme immunoassay for the quantitative measurement of specific IgM antibodies against Gliadin in serum and plasma.
Quantification
Gliadin IgM ELISA Kit
1 Kit
Store the kit at 4°C.
Human
Colorimetric
12 Gliadin Antigen coated microtiter strip, 2mL calibrator A (negative control), 2mL calibrator B (cut-off standard), 2mL calibrator C (weak positive control), 2mL calibrator D (positive control), 15mL Enzyme Conjugate, 15mL Substrate, 15mL stop solution, 60mL sample diluent, 60mL Wash Buffer (10X)
Gliadin IgM ELISA kit
Plasma, Serum
100μL