Learn More
Abnova™ Global Acetyl Histone H3-K18 Quantification Kit (Colorimetric)
Use for measuring global Acetylation of Histone H3-K18
Supplier:  Abnova™ KA0639
Catalog No. 89-027-198
Add to Cart
Description
Description
Specifications
Specifications
Global Acetyl Histone H3-K18 Quantification Kit (Colorimetric) is use for measuring global acetylation of histone H3-K18.
20mL C1 (Wash Buffer 10X), 12mL C2 (Antibody Buffer), 10μL C3 (detection Antibody, 1mg/mL), 10μL C4 (color developer), 6mL C5 (stop solution), 20μL standard control (100μg/mL), 98-wel
Store C3 and standard control at -20°C. Store all other components at 4°C away from light.
Global Acetyl Histone H3-K18 quantification kit
Colorimetric
1 Kit
Quantification