Learn More
Abnova™ Global Acetyl Histone H3-K23 Quantification Kit (Fluorometric)
For measuring global Acetylation of Histone H3-K23
Manufacturer:  Abnova™ KA0642
Catalog No. 89-027-001
Add to cart
Specifications
Specifications
Global Acetyl Histone H3-K23 Quantification Kit (Fluorometric) is use for measuring global acetylation of histone H3-K23.
Quantification
Global Acetyl Histone H3-K23 Quantification Kit (Fluorometric)
1 Kit
Fluorometric
20mL F1 (Wash Buffer 10X), 12mL F2 (Antibody Buffer), 10μL F3 (detection Antibody, 1mg/mL), 24μL F4 (fluoro developer), 24μL F5 (fluoro enhancer), 8mL F6 (fluoro dilution), 20μL stand
Global Acetyl Histone H3-K23 quantification kit
Store F3, F4 and Standard control at -20°C. Store all other components at 4°C away from light.