Learn More
Abnova™ Global Acetyl Histone H4K12 Quantification Kit (Colorimetric)
Used for measuring global acetylation of histone H4K12
Manufacturer:  Abnova™ KA1536
Catalog No. 89-028-230
Add to cart
Specifications
Specifications
20 to 5000ng/well
Quantification
1 Kit
50μL
Colorimetric
Global Acetyl Histone H4K12 Quantification Kit (Colorimetric) is used for measuring global acetylation of histone H4K12.
10mL C1 (wash buffer 10X), 6mL C2 (antibody buffer), 5μL C3 (detection antibody, 1mg/mL), 5mL C4 (color developer), 3mL C5 (stop solution), 5μL signal report solution, 120μL signal enhancer, 10
Histone extract
Store the detection antibody and the standard at -20°C. Store all other components at 4°C away from light.
Global Acetyl Histone H4K12 Quantification Kit