Learn More
Abnova™ Global Acetyl Histone H4K16 Quantification Kit (Colorimetric)
To measure global acetylation of histone H4K16
Manufacturer:  Abnova™ KA1534
Catalog No. 89-028-228
Add to cart
Specifications
Specifications
20 to 5000ng/well
Quantification
1 Kit
50μL
Colorimetric
Global Acetyl Histone H4K16 Quantification Kit (Colorimetric) is used for measuring global acetylation of histone H4K16.
10mL C1 (wash buffer 10X), 6mL C2 (antibody buffer), 5μL C3 (detection antibody, 1mg/mL), 5mL C4 (color developer), 3mL C5 (stop solution), 10μL standard control (100μg/mL), 5μL signa
Histone extract
Store C3 and the standard at -20°C. Store all other components at 4°C away from light.
Global Acetyl Histone H4K16 Quantification Kit